メスピペットピペット、クラスA
30 ml, ±0,03 ml, Black, 530 mm
VWRU10546-032
Borosilicate 3.3 expansion glass, class A.
- Reusable
- Colour coded for easy sorting
Calibrated at 20 °C to deliver. These pipettes are unserialised or batch certified.
Certifications: Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969. Pipettes are colour coded as per ASTM specification E1273.
- Colour code:Black
- 容量:30 ml
- 長さ:530 mm
- Seq:10
- Tolerance:±0,03 ml