粉体漏斗、ガラス、VWR®製
150 mm, 31 mm
Catalog # VWRU10545-912
Supplier: VWR Collection
Jump to:
Product Details & Documents
Made from Type 1 Class A borosilicate 3.3 expansion glass.
- Constructed with a heavy, uniform wall
- Fire-polished rim and stem to provide long service life
- Chemical resistant
- White printing
Funnels comes with short, wide stem, and are useful for adding powdered chemical to reaction vessels.
Certifications: In acc. with ASTM E-1095 standards.
Specifications
- Funnel O-Ø:150 mm
- Seq:04
- Stem O-Ø:31 mm