Orders LinkContactUsLinkComponent
ビーカー、ローフォーム、Griffin
2000 ml, 188 mm, 131 mm, 200 - 2000 ml
  VWRU10754-760
ビーカー、ローフォーム、Griffin
  VWRU10754-760
ビーカー、ローフォーム、Griffin
ビーカー、ローフォーム、Griffin
2000 ml, 188 mm, 131 mm, 200 - 2000 ml
$67.02
1 * 4 items
VWR® low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass. They feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics.
  • Double capacity scale
  • Uniform wall thickness
  • Very good chemical resistance
  • High temperature resistance
Printed with white graduated scale and a durable marking area.
 : Meets ASTM E960, E960-93.
  • 外径:
    131 mm
  • 目盛り範囲:
    200 - 2000 ml
  • 容量:
    2000 ml
  • 高さ:
    188 mm
  • Seq:
    10