VWR®, ステージ、インサート、エクステンション
Holder for slide and Petri dish, Ø: 54 mm
Catalog # 630-2699
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stages, inserts and extensions for inverted microscopes.
Specifications
- 説明:Holder for slide and Petri dish, Ø: 54 mm
- Seq:303