VWR®, バケツライナー、滅菌
Bucket liners for B-7 buckets, sterile, LDPE/nylon, 790×480 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
Bucket liners create an impermeable barrier between the disinfectant and receptacle.
- LDPE/nylon blend
- Irradiated
- Fits B-7 series buckets
Helps ensure the integrity of a disinfectant blend.
包装: Individually bagged, double-bagged in groups of 6. 4 packs of 6 per case.
Specifications
- 説明:Bucket liners for B-7 buckets, sterile
- 長さ×幅:790×480 mm
- Material:LDPE/nylon
- Seq:01