Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
Cylinders 500ml,Graduation Interval 5ml, Graduation Range 25-500 mL 1 * 2 items
VWRU10545-962
:
VWR Collection
Cylinders 500ml,Graduation Interval 5ml, Graduation Range 25-500 mL 1 * 2 items
VWRU10545-962
:
VWR Collection
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, フェイシャルティッシュ、
: VWR Collection
VWR®, ニトリルグローブ
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, ニトリルグローブ、ニトリルライト
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection