Vydac 208TP C8, HPLCカラム
Guard kit for 3,0 – 4,6 mm ID columns, includes 1× guard cartridge, holder and coupler, Guard cartridge, C8, Stainless steel, 300 Å, 3 µm, Silica
Catalog # 208GK34/N
Supplier: HICHROM
Jump to:
Product Details & Documents
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20K MW.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- カラム形式:Guard cartridge
- 粒子サイズ:3 µm
- 細孔サイズ:300 Å
- 説明:Guard kit for 3,0 – 4,6 mm ID columns, includes 1× guard cartridge, holder and coupler
- Seq:44
- Column hardware:Stainless steel
- Phase:C8
- Packing material:Silica
- Cat. No.:208GK34/N
- Environmental certification:ISO 14001:2015 (530840 UM15)
- Environmentally Preferable:Low Manufacturing Impact



