再封可能なバッグ、ZIPPIT®(ジッピット
101,6 µm, 229×305 mm
VWRU89005-310
Plain ZIPPIT® reclosable zipper bags
- Economical
- Can be printed with your logo or instructions
- Including bar code
: In easy-to-use DispenserBag® convenience packaging, designed to function like a 'tissue soft pack'. Pull one bag at a time from the center package opening or open the reclosable zipper at the top to remove.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- 寸法:229×305 mm
- Seq:15
- 厚さ:101,6 µm