マイクロスパチュラ、 両端
Micro spatula with curved, square-cut sharpened tip and rounded spoon at other end, 165 mm, 3 mm
VWRU89259-888
These micro spatulas are double-ended and made of stainless steel.
- 説明:Micro spatula with curved, square-cut sharpened tip and rounded spoon at other end
- Seq:01
- Blade width:3 mm
- Blade length:165 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



