安全キャップ、VWR®
Safety waste cap, including connector for exhaust filter, S 55, 2× for tubing 1,6/3,2 mm OD, 1× for tubing 6,4 - 9 mm OD
Catalog # 590-0045
Supplier: VWR Collection
Jump to:
Product Details & Documents
Collect your solvent waste safely without being exposed to solvent vapours or hazardous gases. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
- Recommended for HPLC systems
- Chemically resistant against aggressive organic solvents
- Tubing connector for e.g. leak connection of diverse HPLC instruments (only for safety waste caps with tube connections)
: Exhaust air filter needs to be ordered separately.
Specifications
- Thread:S 55
- 説明:Safety waste cap, including connector for exhaust filter
- Connector:2× for tubing 1,6/3,2 mm OD, 1× for tubing 6,4 - 9 mm OD