Orders LinkContactUsLinkComponent
安全キャップ、VWR®
Safety waste cap, including connector for exhaust filter, GL 45, 2× for tubing 1,6/3,2 mm OD, 1× for tubing 6,4 - 9 mm OD
  590-0044
安全キャップ、VWR®
  590-0044
安全キャップ、VWR®
安全キャップ、VWR®
Safety waste cap, including connector for exhaust filter, GL 45, 2× for tubing 1,6/3,2 mm OD, 1× for tubing 6,4 - 9 mm OD
1 * 1 items
Collect your solvent waste safely without being exposed to solvent vapours or hazardous gases. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
  • Recommended for HPLC systems
  • Chemically resistant against aggressive organic solvents
  • Tubing connector for e.g. leak connection of diverse HPLC instruments (only for safety waste caps with tube connections)
 : Exhaust air filter needs to be ordered separately.
  • Thread:
    GL 45
  • 説明:
    Safety waste cap, including connector for exhaust filter
  • Connector:
    2× for tubing 1,6/3,2 mm OD, 1× for tubing 6,4 - 9 mm OD