Orders LinkContactUsLinkComponent
粉体漏斗、ガラス、VWR®製
80 mm, 25 mm
  10546-046
粉体漏斗、ガラス、VWR®製
  10546-046
粉体漏斗、ガラス、VWR®製
粉体漏斗、ガラス、VWR®製
80 mm, 25 mm
$159.31
1 * 6 items
Made from Type 1 Class A borosilicate 3.3 expansion glass.
  • Constructed with a heavy, uniform wall
  • Fire-polished rim and stem to provide long service life
  • Chemical resistant
  • White printing
Funnels comes with short, wide stem, and are useful for adding powdered chemical to reaction vessels.
 : In acc. with ASTM E-1095 standards.
  • Funnel O-Ø:
    80 mm
  • Seq:
    02
  • Stem O-Ø:
    25 mm