粉体漏斗、ガラス、VWR®製
80 mm, 25 mm
10546-046
Made from Type 1 Class A borosilicate 3.3 expansion glass.
- Constructed with a heavy, uniform wall
- Fire-polished rim and stem to provide long service life
- Chemical resistant
- White printing
Funnels comes with short, wide stem, and are useful for adding powdered chemical to reaction vessels.
: In acc. with ASTM E-1095 standards.
- Funnel O-Ø:80 mm
- Seq:02
- Stem O-Ø:25 mm