VWR®, 乳棒、細胞ストレイナー用
Pestles, sterile, bulk packaging, Cell strainers, Purple
734-2820
Wear-proof purple polypropylene pestles which are designed to fit into cell strainers perfectly.
- Grooved handle for ease of use
- Sterilised by electron beam irradiation
- Non-pyrogenic, DNase/RNase free
Packaging: Each package has lot number printed on it to facilitate quality tracking. Supplied as individually packaged in easy to tear paper and plastic for easy opening (100/case). Bulk packaging has 25/bag and 50/case.
- For:Cell strainers
- Colour:Purple
- 説明:Pestles, sterile, bulk packaging
- Seq:02
- Cat. No.:734-2820
- Environmental certification:ISO 14001/Certificate No.: 04923E00243R0M
- Environmentally Preferable:Low Manufacturing Impact