Orders LinkContactUsLinkComponent
VWR®, 乳棒、細胞ストレイナー用
VWR®, 乳棒、細胞ストレイナー用
Pestles, sterile, bulk packaging, Cell strainers, Purple
  734-2820
   Low Manufacturing Impact
VWR®, 乳棒、細胞ストレイナー用
  734-2820
   Low Manufacturing Impact
VWR®, 乳棒、細胞ストレイナー用
VWR®, 乳棒、細胞ストレイナー用
Pestles, sterile, bulk packaging, Cell strainers, Purple
1 * 50 items
Wear-proof purple polypropylene pestles which are designed to fit into cell strainers perfectly.
  • Grooved handle for ease of use
  • Sterilised by electron beam irradiation
  • Non-pyrogenic, DNase/RNase free
Packaging: Each package has lot number printed on it to facilitate quality tracking. Supplied as individually packaged in easy to tear paper and plastic for easy opening (100/case). Bulk packaging has 25/bag and 50/case.
  • For:
    Cell strainers
  • Colour:
    Purple
  • 説明:
    Pestles, sterile, bulk packaging
  • Seq:
    02
  • Cat. No.:
    734-2820
  • Environmental certification:
    ISO 14001/Certificate No.: 04923E00243R0M
  • Environmentally Preferable:
    Low Manufacturing Impact