VWR®, 乳棒、細胞ストレイナー用
Pestle, sterile, individually packaged, Cell strainers, Purple
Catalog # 734-2819
Supplier: VWR Collection
Jump to:
Product Details & Documents
Wear-proof purple polypropylene pestles which are designed to fit into cell strainers perfectly.
- Grooved handle for ease of use
- Sterilised by electron beam irradiation
- Non-pyrogenic, DNase/RNase free
包装: Each package has lot number printed on it to facilitate quality tracking. Supplied as individually packaged in easy to tear paper and plastic for easy opening (100/case). Bulk packaging has 25/bag and 50/case.
Specifications
- For:Cell strainers
- Colour:Purple
- 説明:Pestle, sterile, individually packaged
- Seq:01
- Cat. No.:734-2819
- Environmental certification:ISO 14001/Certificate No.: 04923E00243R0M
- Environmentally Preferable:Low Manufacturing Impact