Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, カーボーイ、丸型, サニタリーネック, PP

VWR®, カーボーイ、丸型, サニタリーネック, PP

10 L, 76 mm
Catalog # 216-1920
Supplier:   VWR Collection
VWR®, カーボーイ、丸型, サニタリーネック, PP
Catalog # 216-1920
Supplier:  VWR Collection
VWR®, カーボーイ、丸型, サニタリーネック, PP
VWR®, カーボーイ、丸型, サニタリーネック, PP

10 L, 76 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Ideal for bioprocessing applications. Features a sanitary flange (76 mm) that accepts standard TC fittings.

  • Convenient shoulder handles
  • Very good chemical resistance
  • Autoclavable

Certifications: Resin meets US FDA 21 CFR, USP Class VI.

Documents

Specifications
  • 容量:
    10 L
  • Seq:
    01
  • Neck-Ø:
    76 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.