Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, カーボーイ、丸型, サニタリーネック, PP

20 L, 76 mm
  216-1897
VWR®, カーボーイ、丸型, サニタリーネック, PP
  216-1897
VWR®, カーボーイ、丸型, サニタリーネック, PP
VWR®, カーボーイ、丸型, サニタリーネック, PP

20 L, 76 mm

 
 
   
1 * 1 items
 

Ideal for bioprocessing applications. Features a sanitary flange (76 mm) that accepts standard TC fittings.

  • Convenient shoulder handles
  • Very good chemical resistance
  • Autoclavable

Certifications: Resin meets US FDA 21 CFR, USP Class VI.

 
  • 容量:
    20 L
  • Seq:
    03
  • Neck-Ø:
    76 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.