メスピペットピペット、クラスA
1 ml, ±0,006 ml, Blue, 315 mm
Catalog # VWRU10546-006
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 expansion glass, class A.
- Reusable
- Colour coded for easy sorting
Calibrated at 20 °C to deliver. These pipettes are unserialised or batch certified.
: Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969. Pipettes are colour coded as per ASTM specification E1273.
Specifications
- Colour code:Blue
- 容量:1 ml
- 長さ:315 mm
- Seq:01
- Tolerance:±0,006 ml