VWR®カーボーイ用スペアキャップ
Cap for narrow mouth VWR carboys, 83 mm, Purple
Catalog # 216-1840
Supplier: VWR Collection
Jump to:
Product Details & Documents
Spare caps are designed for use with either narrow mouth or wide mouth VWR® carboys
Autoclavable.
Documents
Avantor Cell Culture Solutions.pdf
Product Literature
CoC VWR Carboys Narrow Neck Round with Stopcock and Screw Cap CoC VWR Carboys with Screw Cap, Wide Neck, Round CoC VWR Round Carboys with Narrow Neck and Purple Screw Caps CoC VWR®, Carboys, Narrow Neck, Heavy-Duty, Round, with Screw Cap
TDS VWR Carboys.pdf
Datasheet
Specifications
- キャップサイズ:83 mm
- Colour:Purple
- 説明:Cap for narrow mouth VWR carboys
- Seq:01