VWR®, 滴下瓶、
Pipette kit for 10 and 15 ml dropping bottles, -
Catalog # 215-3824
Supplier: VWR Collection
Jump to:
Product Details & Documents
Soda-lime glass, amber.
- Good chemical and physical properties
- Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Specifications
- 容量:-
- 説明:Pipette kit for 10 and 15 ml dropping bottles
- Seq:09