Orders LinkContactUsLinkComponent
VWR®, 滴下瓶、
VWR®, 滴下瓶、
Bottle only, 50 ml
Catalog # 215-3821
Supplier:   VWR Collection
VWR®, 滴下瓶、
Catalog # 215-3821
Supplier:  VWR Collection
VWR®, 滴下瓶、
VWR®, 滴下瓶、
Bottle only, 50 ml
Price
Inquire For Price
Log in for availability
1 * 144 items
Product Details & Documents
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Specifications
  • 容量:
    50 ml
  • 説明:
    Bottle only
  • Seq:
    06