VWR®, 滴下瓶、
Bottle only, 5 ml
Catalog # 215-3816
Supplier: VWR Collection
Jump to:
Product Details & Documents
Soda-lime glass, amber.
- Good chemical and physical properties
- Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Specifications
- 容量:5 ml
- 説明:Bottle only
- Seq:01