VWR®, マイクロチューブ収納ボックス、
81, Black, 130×130×47 mm
Catalog # 525-0941
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP.
- Highly transparent, with bold identifying colours and a distinctive wave design
- Uniform wells with location ID; capacity indicated on the outside of the box
- Black light-tight version available for light-sensitive samples
- Frosted labelling patch
- Alphanumeric identification on the bottom
Compact footprint for microtube and cryogenic vial storage. Store 1,5/2,0 ml microtubes or cryogenic vials. Complete with an easy open, friction-fit lid. Will fit into most standard freezer racking.
納品情報: Assorted pack contains one of each colour blue, green, pink, yellow and orange.
Specifications
- Compartments:81
- W×D×H ext.:130×130×47 mm
- Colour:Black
- Seq:10