Orders LinkContactUsLinkComponent
VWR®, マイクロチューブ収納ボックス、
VWR®, マイクロチューブ収納ボックス、
100, Natural, 141×151×47 mm
Catalog # 525-0926
Supplier:   VWR Collection
VWR®, マイクロチューブ収納ボックス、
Catalog # 525-0926
Supplier:  VWR Collection
VWR®, マイクロチューブ収納ボックス、
VWR®, マイクロチューブ収納ボックス、
100, Natural, 141×151×47 mm
Price
Inquire For Price
Log in for availability
1 * 5 items
Product Details & Documents
PP.
  • Highly transparent, with bold identifying colours and a distinctive wave design
  • Uniform wells with location ID; capacity indicated on the outside of the box
  • Black light-tight version available for light-sensitive samples
  • Frosted labelling patch
  • Alphanumeric identification on the bottom
Compact footprint for microtube and cryogenic vial storage. Store 1,5/2,0 ml microtubes or cryogenic vials. Complete with an easy open, friction-fit lid. Will fit into most standard freezer racking.
納品情報: Assorted pack contains one of each colour blue, green, pink, yellow and orange.
Specifications
  • Compartments:
    100
  • W×D×H ext.:
    141×151×47 mm
  • Colour:
    Natural
  • Seq:
    12