Orders LinkContactUsLinkComponent
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り
25 (for mini bo×), 67×67×32 mm
Catalog # VWRU89214-740
Supplier:   VWR Collection
VWR®, クライオボックスおよび仕切り
Catalog # VWRU89214-740
Supplier:  VWR Collection
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り
25 (for mini bo×), 67×67×32 mm
Price
$4.82
Log in for availability
1 * 1 items
Product Details & Documents
Cardboard, white.
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.
Specifications
  • Compartments:
    25 (for mini bo×)
  • W×D×H ext.:
    67×67×32 mm
  • Seq:
    13