VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
Product Details & Documents
Cleanroom paper is designed for use in cleanroom environments, where the prevention of chemical and particle contamination is critical.
- High tensile strength, and excellent heat resistance, for enhanced copier and printing efficiency
- PE-binding technology results in very low levels of particles, non volatile residue and ion extractables
- Unique precision-cutting technology guarantees consistent sizing and cleanliness
- Paper is web-cleaned on both sides prior to sheeting
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double-bagged in an ISO Class 5 (FED-STD-209E Class 100) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10) to ISO Class 6 (FED-STD-209E Class 1000) cleanrooms.
Poly paper excels in printability. Synthetic paper excels in tensile strength.
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double bagged in an ISO Class 5 (FED-STD-209E Class 100/M3,5) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2,5) to ISO Class 6 (FED-STD-209E Class 1000/M4,5) cleanrooms.
包装: Paper is packed 250 sheets per pack, 10 packs per case.
Documents
Avantor Pureimage Cleanroom Paper and Notebooks
Product Literature
Specifications
- Weight:104 g/m²
- Colour:White
- 説明:Latex-free paper, A4 size, synthetic substrate
- Seq:20
- Cat. No.:115-1957
- Environmentally Preferable:Sustainable Packaging