Orders LinkContactUsLinkComponent
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
Latex-free paper, A3 size, synthetic substrate, Blue, 104 g/m²
Catalog # 115-1954
   Sustainable Packaging
Supplier:   VWR Collection
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
Catalog # 115-1954
Supplier:  VWR Collection
   Sustainable Packaging
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
VWR® PUREImage, クリーンルーム用ペーパー, ピュアイメージ
Latex-free paper, A3 size, synthetic substrate, Blue, 104 g/m²
Price
Inquire For Price
Log in for availability
1 * 1.250 items
Product Details & Documents
Cleanroom paper is designed for use in cleanroom environments, where the prevention of chemical and particle contamination is critical.
  • High tensile strength, and excellent heat resistance, for enhanced copier and printing efficiency
  • PE-binding technology results in very low levels of particles, non volatile residue and ion extractables
  • Unique precision-cutting technology guarantees consistent sizing and cleanliness
  • Paper is web-cleaned on both sides prior to sheeting
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double-bagged in an ISO Class 5 (FED-STD-209E Class 100) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10) to ISO Class 6 (FED-STD-209E Class 1000) cleanrooms.
Poly paper excels in printability. Synthetic paper excels in tensile strength.
Produced in an ISO Class 4 (FED-STD-209E Class 10/M2,5) cleanroom and cleaned and double bagged in an ISO Class 5 (FED-STD-209E Class 100/M3,5) cleanroom. Paper is suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2,5) to ISO Class 6 (FED-STD-209E Class 1000/M4,5) cleanrooms.
 : Paper is packed 250 sheets per pack, 10 packs per case.
Specifications
  • Weight:
    104 g/m²
  • Colour:
    Blue
  • 説明:
    Latex-free paper, A3 size, synthetic substrate
  • Seq:
    11
  • Cat. No.:
    115-1954
  • Environmentally Preferable:
    Sustainable Packaging