VWR®, 絶縁ガウン
White, 1150×1360 mm
These disposable gowns, with back fastening, are made of non woven, breathable SMS fabric. Ideal for use in healthcare environments and hospitals.
- Long sleeves, knitted cuffs
- Round neck
- Ties on neck and waist
- Latex-free
Weight: 45 or 60 g/m²
CoC VWR® Isolation Gowns coc_VWRI113-8315_15220240 coc_VWRI113-8315_19120256 coc_VWRI113-8315_21120251 COC_VWRI113-8315_30920217
- Colour:White
- W×L:1150×1360 mm
- Cat. No.:113-8315
- Garment application:Medical|Clinical
- Gender:Unisex
- Closure type:Back fastening
- Weight:45 g/m²