Orders LinkContactUsLinkComponent
VWR®, オーバーオール、SPP
White, XL
  113-8313
VWR®, オーバーオール、SPP
  113-8313
VWR®, オーバーオール、SPP
VWR®, オーバーオール、SPP
White, XL
1 * 25 items
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • With collar
  • Two-way nylon zip
  • Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
  • 衣服のサイズ:
    XL
  • Colour:
    White
  • Seq:
    03
  • Cat. No.:
    113-8313
  • Model:
    With collar
  • Garment application:
    General Purpose|Industrial
  • Weight:
    40 g/m²