Orders LinkContactUsLinkComponent
VWR®, ESDオーバーオール、フード付き
VWR®, ESDオーバーオール、フード付き
White, 4XL
Catalog # 113-8310
Supplier:   VWR Collection
VWR®, ESDオーバーオール、フード付き
Catalog # 113-8310
Supplier:  VWR Collection
VWR®, ESDオーバーオール、フード付き
VWR®, ESDオーバーオール、フード付き
White, 4XL
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
These breathable overalls are made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Elasticated hood
  • Zip closure
  • Elasticated cuffs, ankles and waist
  • Anti-static properties
Weight: 50 g/m²
Colour: White
 : 1 piece per bag, 50 bags per case.
Specifications
  • 衣服のサイズ:
    4XL
  • Colour:
    White
  • Seq:
    06
  • Cat. No.:
    113-8310
  • Cleanroom class:
    100/ISO 5
  • Anti-static:
    +
  • Garment application:
    General Purpose|Industrial
  • Weight:
    50 g/m²