VWR®, ESDオーバーオール、フード付き
White, 3XL
Catalog # 113-8309
Supplier: VWR Collection
Jump to:
Product Details & Documents
These breathable overalls are made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Elasticated hood
- Zip closure
- Elasticated cuffs, ankles and waist
- Anti-static properties
Weight: 50 g/m²
Colour: White
包装: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
TDS ESD Overalls, SPP, With Hood
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- 衣服のサイズ:3XL
- Colour:White
- Seq:05
- Cat. No.:113-8309
- Cleanroom class:100/ISO 5
- Anti-static:+
- Garment application:General Purpose|Industrial
- Weight:50 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



