VWR®, ESDオーバーオール、フード付き
White, XL
Catalog # 113-8307
Supplier: VWR Collection
Jump to:
Product Details & Documents
These breathable overalls are made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Elasticated hood
- Zip closure
- Elasticated cuffs, ankles and waist
- Anti-static properties
Weight: 50 g/m²
Colour: White
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
TDS ESD Overalls, SPP, With Hood
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- 衣服のサイズ:XL
- Colour:White
- Seq:03
- Cat. No.:113-8307
- Cleanroom class:100/ISO 5
- Anti-static:+
- Garment application:General Purpose|Industrial
- Weight:50 g/m²