Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 実験用白衣、SPP、使い捨て

VWR®, 実験用白衣、SPP、使い捨て

SPP 45 g/m², White, 4XL
Catalog # 113-8304
Supplier:   VWR Collection
VWR®, 実験用白衣、SPP、使い捨て
Catalog # 113-8304
Supplier:  VWR Collection
VWR®, 実験用白衣、SPP、使い捨て
VWR®, 実験用白衣、SPP、使い捨て

SPP 45 g/m², White, 4XL

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

This disposable coat is made of non woven, laminated, SPP fabric.

  • Tailored collar
  • Press stud closure and open cuffs
  • Two pockets

Suitable for use in ISO Class 5 + 6 cleanroom environments.

Weight: 45/30 g/m²

Specifications
  • 衣服のサイズ:
    4XL
  • Colour:
    White
  • 説明:
    SPP 45 g/m²
  • Seq:
    06
  • Cat. No.:
    113-8304
  • Garment application:
    General Purpose|Laboratory
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Buttons