Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 実験用白衣、SPP、使い捨て

VWR®, 実験用白衣、SPP、使い捨て

SPP 45 g/m², White, 3XL
Catalog # 113-8303
Supplier:   VWR Collection
VWR®, 実験用白衣、SPP、使い捨て
Catalog # 113-8303
Supplier:  VWR Collection
VWR®, 実験用白衣、SPP、使い捨て
VWR®, 実験用白衣、SPP、使い捨て

SPP 45 g/m², White, 3XL

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

This disposable coat is made of non woven, laminated, SPP fabric.

  • Tailored collar
  • Press stud closure and open cuffs
  • Two pockets

Suitable for use in ISO Class 5 + 6 cleanroom environments.

Weight: 45/30 g/m²

Specifications
  • 衣服のサイズ:
    3XL
  • Colour:
    White
  • 説明:
    SPP 45 g/m²
  • Seq:
    05
  • Cat. No.:
    113-8303
  • Garment application:
    General Purpose|Laboratory
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Buttons
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.