Orders LinkContactUsLinkComponent
VWR®, オーバーオール、PP、フード付き
VWR®, オーバーオール、PP、フード付き
White, L
Catalog # 113-8262
Supplier:   VWR Collection
VWR®, オーバーオール、PP、フード付き
Catalog # 113-8262
Supplier:  VWR Collection
VWR®, オーバーオール、PP、フード付き
VWR®, オーバーオール、PP、フード付き
White, L
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
Overalls made from non woven PP. Ideal for non critical applications.
  • Hood, long sleeves and legs
  • Elasticated waist, wrist and ankles
  • Bound seams and zip fastening
Weight: 40 g/m²
Colour: White
Specifications
  • 衣服のサイズ:
    L
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    113-8262
  • Garment application:
    General Purpose|Industrial
  • Weight:
    40 g/m²