VWR®, オーバーオール、PP、フード付き
White, L
Supplier: VWR Collection
Jump to:
Product Details & Documents
Overalls made from non woven PP. Ideal for non critical applications.
- Hood, long sleeves and legs
- Elasticated waist, wrist and ankles
- Bound seams and zip fastening
Weight: 40 g/m²
Colour: White
Specifications
- 衣服のサイズ:L
- Colour:White
- Seq:02
- Cat. No.:113-8262
- Garment application:General Purpose|Industrial
- Weight:40 g/m²