VWR®, オーバーオール、SMS
Product Details & Documents
These durable, anti-static overalls are made of breathable, soft SMS, a spunbonded fabric with a unique tri-laminate construction, that offers a high tensile strength and toughness, plus provides a barrier to fluids and particulates.
- Elasticated hood
- Zip closure with flap
- Elasticated cuffs, ankles and waist, thumb loop
- Serged seams (stitching method)
- Low lint performance
These overalls can be used in medical, industrial, laboratory, electronic and pharmaceutical applications. Suitable for use in Critical Environments (IEST-RP-CC003.3 – Level I) and ISO Class 5 + 6 cleanrooms.
Weight: 60 g/m²
EN 340, EN 1149-5, EN 13034:2005+A1:2009 (Type 5), EN 13982-1:2004+A1:2010 (Type 6), EN 1073-2:2002; Cat. III
Certifications: EN 340, EN 1149-5, EN 13034+A1 (Type 5), EN 13982-1+A1 (Type 6), EN 1073-2; Cat. III
Documents
CoC Coverall CAT III Type 5,6.pdf
Conformity declaration
Specifications
- 衣服のサイズ:L
- Colour:White
- Seq:03
- Cat. No.:113-8227
- Anti-static:+
- Garment application:Cleanroom|Medical|Laboratory|Clinical|Industrial
- Weight:60 g/m²