VWR®, 保護フードとフェースカバー
Hood and face cover, White, L
These poypropylene head and face covers are designed to protect users from potential environmental contaminations.
- Bulk packaging
- White colour
Weight: 20 g
: Supplied in a case of 500, garments are bulk packed with polybag and case liner.
- 衣服のサイズ:L
- Colour:White
- 説明:Hood and face cover
- Seq:01
- Cat. No.:113-8202
- Garment application:Laboratory