Orders LinkContactUsLinkComponent
VWR®, 保護フードとフェースカバー
Hood and face cover, White, L
  113-8202
VWR®, 保護フードとフェースカバー
  113-8202
VWR®, 保護フードとフェースカバー
VWR®, 保護フードとフェースカバー
Hood and face cover, White, L
1 * 500 items
These poypropylene head and face covers are designed to protect users from potential environmental contaminations.
  • Bulk packaging
  • White colour
Weight: 20 g
 : Supplied in a case of 500, garments are bulk packed with polybag and case liner.
  • 衣服のサイズ:
    L
  • Colour:
    White
  • 説明:
    Hood and face cover
  • Seq:
    01
  • Cat. No.:
    113-8202
  • Garment application:
    Laboratory