クリーンルーム用スパイラルノート、 VWR®
Cleanroom spiral notebook, latex, Side-spiral, 140×216 mm
Catalog # VWRU12345-040
Supplier: VWR Collection
Jump to:
Product Details & Documents
These spiral-bound notebooks feature latex-free paper in college-rule style.
- Semi-transparent, HDPE cover
- Fully recyclable pages
Notebooks have 100 pages (50 sheets) each.
Specifications
- Paper size:140×216 mm
- Type:Side-spiral
- 説明:Cleanroom spiral notebook, latex
- Seq:02