Order Entry
Japan
Orders LinkContactUsLinkComponent
 

マイクロスパチュラ、 両端

Micro spatula with bent, sharpened tip and V-shaped sharpened spoon at other end, 170 mm, 2 mm
Catalog # VWRU89259-890
Supplier:   VWR Collection
マイクロスパチュラ、 両端
Catalog # VWRU89259-890
Supplier:  VWR Collection
マイクロスパチュラ、 両端
マイクロスパチュラ、 両端

Micro spatula with bent, sharpened tip and V-shaped sharpened spoon at other end, 170 mm, 2 mm

Price
$18.08
Log in for availability
1 * 1 items
Product Details & Documents

These micro spatulas are double-ended and made of stainless steel.

Specifications
  • 説明:
    Micro spatula with bent, sharpened tip and V-shaped sharpened spoon at other end
  • Seq:
    02
  • Blade width:
    2 mm
  • Blade length:
    170 mm