微量遠沈管、1.7/2.0 ml
2,0 ml, Colourless, with attached lid, graduated, conical bottom, non sterile, -
Catalog # VWRU87003-298
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP, graduated, autoclavable and freezable with attached flat caps.
- DNase-, RNase and endotoxin-free
- Can withstand centrifugation to RCF 20000×g
- Fit all standard and high capacity rotors
- Tubes with X-Resin™ provides ultra-low binding characteristics
Medical grade superior clarity for visualisation of samples and isolated pellets. Graduated frosted side panel and flat snap cap provide surface for labelling.
Specifications
- Sterile:-
- Type:with attached lid
- Version:graduated, conical bottom, non sterile
- 容量:2,0 ml
- Colour:Colourless