Order Entry
Japan
Orders LinkContactUsLinkComponent
 

デシケーターキャビネット、アクリル製、VWR®

Desiccator cabinet, clear acrylic, 23,2 L, 3, 305×305×305 mm
Catalog # VWRU32800-012
Supplier:   VWR Collection
デシケーターキャビネット、アクリル製、VWR®
Catalog # VWRU32800-012
Supplier:  VWR Collection
デシケーターキャビネット、アクリル製、VWR®
デシケーターキャビネット、アクリル製、VWR®

Desiccator cabinet, clear acrylic, 23,2 L, 3, 305×305×305 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These acrylic desiccator cabinets can be used in refrigerators or at room temperature for dry storage of moisture-sensitive materials.

  • Fully removable, tray-like shelves have perforated bottoms and slide in/out on pre-set rails
  • Door opens from right to left and is secured with a stainless steel latch

Acrylic desiccator cabinets are fabricated from either clear or bronze acrylic, with a variable number of trays depending on model. Clear acrylic for easy viewing or bronze for light-sensitive materials. Bronze acrylic features stainless steel twist locking latches and hinges.

Specifications
  • W×D×H ext.:
    305×305×305 mm
  • 説明:
    Desiccator cabinet, clear acrylic, 23,2 L
  • Seq:
    03
  • Shelves:
    3