VWR®, Tissue Processing/Embedding Cassettes, Premium
Tissue cassettes, Premium, Turquoise
Catalog # VWRU18000-004
Supplier: VWR Collection
VWR®, Tissue Processing/Embedding Cassettes, Premium
Catalog # VWRU18000-004
Supplier: VWR Collection
Jump to:
Product Details & Documents
These cassettes made of acetyl polymer feature a fluidic flow design for the optimisation of tissue processing and embedding.
- Resistant to histological solvents
- Chamber is 10% longer allowing for an increased specimen range
- 45° angled print surface compatible with all automatic printing devices
- Attached cover has a large labelling area and uses a frangible hinge
- Unique design with vented side walls to maximise fluid exchange
The vented sides and rear wall design work exceptionally well with fatty, dense, and other difficult to process tissue samples.
Specifications
- Colour:Turquoise
- 説明:Tissue cassettes, Premium
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



