VWR®, バッグ、バイオハザード
PP, 60 L, 914×610 mm, 100 µm
Catalog # VWRU14221-994
Supplier: VWR Collection
Jump to:
Product Details & Documents
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 75 µm thick, orange, with 'Biohazard' label.
- Autoclavable up to 135 °C
- The useful temperature indicator text is white before autoclaving, then black
- 'Contents Have Been Autoclaved' indicates successful autoclaving
- Luminous red for easy recognition and highly visible 'Biohazard' label with the corresponding warning sign
These bags are DOT approved.
Specifications
- 容量:60 L
- 長さ×幅:914×610 mm
- Material:PP
- Seq:10
- 厚さ:100 µm



