Orders LinkContactUsLinkComponent
ビーカー、低形状、PP
ビーカー、低形状、PP
100 ml, 10 - 100 ml
Catalog # VWRU13890-046
Supplier:   VWR Collection
ビーカー、低形状、PP
Catalog # VWRU13890-046
Supplier:  VWR Collection
ビーカー、低形状、PP
ビーカー、低形状、PP
100 ml, 10 - 100 ml
Price
$75.29
Log in for availability
1 * 12 items
Product Details & Documents
PP, translucent, graduated.
  • Features no-drip spout for clean pouring
  • Tapered walls for secure grip
  • With an oval marking spot
Ideal for general laboratory procedures. These are easy-to-handle stackable beakers and is break-resistant, nontoxic, non-contaminating, and extremely chemical-resistant. Sturdy construction can withstand repeated autoclaving.
Specifications
  • 目盛り範囲:
    10 - 100 ml
  • 容量:
    100 ml
  • Seq:
    02