VWR®, ピペットチップ、スタンダード
0,5 - 20 µl, -, Transparent, Bulk
Catalog # 613-3019
Supplier: VWR Collection
Jump to:
Product Details & Documents
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
Ordering information: Supplied loose or in racks holding 96 tips.
Specifications
- Packed:Bulk
- Sterile:-
- 容量:0,5 - 20 µl
- Seq:2
- Tip colour:Transparent