Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ラック、VLSBシェーキングバス用、

VWR®, ラック、VLSBシェーキングバス用、

Stainless steel test tube rack for 12 and 18 l baths, 10, 30 mm
Catalog # 462-0507
Supplier:   VWR Collection
VWR®, ラック、VLSBシェーキングバス用、
Catalog # 462-0507
Supplier:  VWR Collection
VWR®, ラック、VLSBシェーキングバス用、
VWR®, ラック、VLSBシェーキングバス用、

Stainless steel test tube rack for 12 and 18 l baths, 10, 30 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • For tube Ø:
    30 mm
  • 穴の数:
    10
  • 説明:
    Stainless steel test tube rack for 12 and 18 l baths
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.