Orders LinkContactUsLinkComponent
微量遠沈管、1.7/2.0 ml
微量遠沈管、1.7/2.0 ml
1,7 ml, Colourless, with attached lid, graduated, conical bottom, non sterile, -
Catalog # VWRU87003-294
Supplier:   VWR Collection
微量遠沈管、1.7/2.0 ml
Catalog # VWRU87003-294
Supplier:  VWR Collection
微量遠沈管、1.7/2.0 ml
1,7 ml, Colourless, with attached lid, graduated, conical bottom, non sterile, -
Price
$33.87
Log in for availability
1 * 500 items
Product Details & Documents
PP, graduated, autoclavable and freezable with attached flat caps.
  • DNase-, RNase and endotoxin-free
  • Can withstand centrifugation to RCF 20000×g
  • Fit all standard and high capacity rotors
  • Tubes with X-Resin™ provides ultra-low binding characteristics
Medical grade superior clarity for visualisation of samples and isolated pellets. Graduated frosted side panel and flat snap cap provide surface for labelling.
Specifications
  • Sterile:
    -
  • Type:
    with attached lid
  • Version:
    graduated, conical bottom, non sterile
  • 容量:
    1,7 ml
  • Colour:
    Colourless