Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GC-rich DNA Target Kit: TEMPase Hot Start 5u/µl 50r, 4x GC buffer I, 4x GC buffer II, 25 mM MgCl2 1 * 1 Each
Catalog # 733-2332SAMPLE
Supplier:
VWR Chemicals
Expand image
GC-rich DNA Target Kit: TEMPase Hot Start 5u/µl 50r, 4x GC buffer I, 4x GC buffer II, 25 mM MgCl2 1 * 1 Each
Catalog # 733-2332SAMPLE
Supplier:
VWR Chemicals
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, ニトリルグローブ、ニトリルライト
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, ニトリルグローブ
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, フェイシャルティッシュ、
Supplier: VWR Collection