Order Entry
Japan
Orders LinkContactUsLinkComponent
PCRバッファー

PCRバッファー

10X Extra Buffer, 15 mM MgCl₂, 3× 1,5 ml, 1 KIT
Catalog # 733-2303
Supplier:   VWR Chemicals
PCRバッファー
Catalog # 733-2303
Supplier:  VWR Chemicals
CAS Number:  
PCRバッファー
PCRバッファー

10X Extra Buffer, 15 mM MgCl₂, 3× 1,5 ml, 1 KIT

CAS Number:  
Price
$82.74
Log in for availability
1 * 1 KIT
Product Details & Documents

An optimal buffer system is critical for the performance of successful PCR. VWR® Taq DNA Polymerase kits generally include two different buffers, Key Buffer and Extra Buffer, which are suited for different PCR needs. All buffers contain Tris and 15 mM MgCl₂ (1,5 mM MgCl₂ final concentration). Additional MgCl₂ for easy optimisation is included in a separate vial.

Key Buffer: Key Buffer (NH⁴⁺) gives a superior amplification signal (high yield) and minimises the need for optimisation of the Mg²⁺ concentration or the annealing temperature in most primer-template systems.

Extra Buffer: Extra Buffer is the traditional potassium (K+) buffer. Extra Buffer promotes high specificity. Careful optimisation of primer annealing temperatures and Mg²⁺ concentrations may be required.

Combination Buffer: Combination Buffer is a proprietary mixture of K+ and NH⁴⁺. It combines high specificity with good product yield and high tolerance to optimisation of primer annealing temperatures and Mg²⁺ concentrations due to its balanced ammonium-potassium formulation.

Buffers for GC-rich templates: Combined with VWR® TEMPase the GC buffers promote excellent amplification results with targets of varying degrees of GC content. This confirms that the choice of buffer is crucial for any successful amplification.

Specifications

Specifications

  • Pk:
    1 KIT
  • 説明:
    10X Extra Buffer, 15 mM MgCl₂
  • 容量:
    3× 1,5 ml
  • Cat. No.:
    733-2303
  • Description:
    Extra Buffer
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Size:
    3 x 1.5 ml
  • Concentration:
    10x
  • Application:
    Standard amplification

Specifications