Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 乳鉢、乳棒付き、浅型

VWR®, 乳鉢、乳棒付き、浅型

114 mm, 127 mm
Catalog # VWRU89037-494
Supplier:   VWR Collection
VWR®, 乳鉢、乳棒付き、浅型
Catalog # VWRU89037-494
Supplier:  VWR Collection
VWR®, 乳鉢、乳棒付き、浅型
VWR®, 乳鉢、乳棒付き、浅型

114 mm, 127 mm

Price
$1,242.79
Log in for availability
1 * 1 items
Product Details & Documents

Agate.

  • High quality with flawless grinding surface
Specifications
  • 直径:
    127 mm
  • 内径:
    114 mm
  • Seq:
    12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.