J.T.Baker®, Hematology Controls, 3-diff Control Extended
3-Different control extended, high, 2,5 mL
Supplier: Avantor
J.T.Baker®, Hematology Controls, 3-diff Control Extended
Catalog # 3423.
Supplier: Avantor
CAS Number:
Jump to:
Product Details & Documents
In vitro diagnostic hematology control product, intended for professional use only.
Caution: Parameter Controls are to be stored upright at 2-8°C when not in use. Stored at this temperature, the Controls are guaranteed stable until the expiry date.
Documents
Specifications
Specifications
- Pk:2,5 mL
- 説明:3-Different control extended, high